Learn More
Wl Gore And Associates 1.0 mL GORE Protein Capture Device with Protein A for Rapid mAb Purification
Catalog No. NC1267631
Encompass
GORE Protein Capture Devices with immobilized Protein A are intended for small-scale affinity purification of monoclonal antibodies and other proteins demonstrating an Fc region. GORE Protein Capture Devices use a unique expanded polytetrafluoroethylene (ePTFE) membrane composite that provides a binding capacity advantage at high flow rates and improved the speed of purification. In addition the GORE Protein Capture Deivces produce highly concentrated elution pools, which may eliminate a downstream concentration step during the antibody purification process. These unique membrane devices are offered in 1 mL (≥ 30 mg capacity), 3.5 mL (≥ 105 mg capacity) and 9.0 mL (≥27 mg capacity) at 20 seconds residence time.
Catalog No. NC1267631 Supplier Wl Gore And Associates Supplier No. PROA101
GORE Protein Capture Devices with immobilized Protein A are intended for small-scale affinity purification of monoclonal antibodies and other proteins demonstrating an Fc region. GORE Protein Capture Devices use a unique expanded polytetrafluoroethylene (ePTFE) membrane composite that provides a binding capacity advantage at high flow rates and improved the speed of purification. In addition the GORE Protein Capture Deivces produce highly concentrated elution pools, which may eliminate a downstream concentration step during the antibody purification process. These unique membrane devices are offered in 1 mL (≥ 30 mg capacity), 3.5 mL (≥ 105 mg capacity) and 9.0 mL (≥27 mg capacity) at 20 seconds residence time.