Learn More
Chemglass Life Sciences 1.0mL 96 Well Multi-Tier Reaction Plate, Topas, Complete
SDP
Catalog No. 50872348
Encompass_Preferred
1.0mL 96 Well Multi-Tier Reaction Plate, Topas, Complete. Our Multi-Tier 96 well reaction plates have a fixed height or can be multi-tiered to accept up to 1.5mL borosilicate glass inserts. Plates have the same footprint as standard 96 well polystyrene plates and are compatible with 96 well automation workstations. Complete setups include: Base Plate, Glass Inserts, Prescored PTFE/Silicone Septa, and Snap On Cover. PLEASE NOTE: PLATES ARE AUTOCLAVABLE.
Catalog No. 50-872-348 Supplier Chemglass Life Sciences Supplier No. CG191502
Add to Cart
Add to Cart
1.0mL 96 Well Multi-Tier Reaction Plate, Topas, Complete. Our Multi-Tier 96 well reaction plates have a fixed height or can be multi-tiered to accept up to 1.5mL borosilicate glass inserts. Plates have the same footprint as standard 96 well polystyrene plates and are compatible with 96 well automation workstations. Complete setups include: Base Plate, Glass Inserts, Prescored PTFE/Silicone Septa, and Snap On Cover. PLEASE NOTE: PLATES ARE AUTOCLAVABLE.