Learn More
ANALYTIK JENA US LLC 1.0MM Quartz Glass Injector, Minimized Clogging, High Resolution Optics, Unit
Catalog No. NC1727867
Encompass
The 1.0 mm quartz glass injector is essential for optimal performance in Analytik Jena's PlasmaQuant 9100 ICP-OES systems. This injector enhances plasma stability and minimizes clogging and soot formation, ensuring reliable trace element analysis. Its robust design offers compatibility with the V Shuttle Torch, making it suitable for challenging samples including volatile organics. With benefits like reduced maintenance needs and high-resolution optics support, it stands out for its versatility and superior analytical performance.
Catalog No. NC1727867 Supplier ANALYTIK JENA US LLC Supplier No. 41813410408
The 1.0 mm quartz glass injector is essential for optimal performance in Analytik Jena's PlasmaQuant 9100 ICP-OES systems. This injector enhances plasma stability and minimizes clogging and soot formation, ensuring reliable trace element analysis. Its robust design offers compatibility with the V Shuttle Torch, making it suitable for challenging samples including volatile organics. With benefits like reduced maintenance needs and high-resolution optics support, it stands out for its versatility and superior analytical performance.