Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metrohm USA 1.0N H2SO4, 1 L, 6 bottles, Case
SDP

Catalog No. NC0441429 Shop All Metrohm USA Products
Encompass

This precisely formulated sulfuric acid solution from Metrohm USA is designed for diverse scientific applications. It is a reliable reagent used in various analytical and preparative laboratory procedures, making it essential for any laboratory setting.

  • Chemical name: Sulfuric acid
  • Chemical formula: H2SO4
  • Concentration: 1.0 N
  • Unit volume: 1 L
  • Units per case: 6 bottles
  • Packaging: Case
  • Application: General laboratory reagent

Catalog No. NC0441429 Supplier Metrohm USA Supplier No. LABTI90701
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This precisely formulated sulfuric acid solution from Metrohm USA is designed for diverse scientific applications. It is a reliable reagent used in various analytical and preparative laboratory procedures, making it essential for any laboratory setting.

  • Chemical name: Sulfuric acid
  • Chemical formula: H2SO4
  • Concentration: 1.0 N
  • Unit volume: 1 L
  • Units per case: 6 bottles
  • Packaging: Case
  • Application: General laboratory reagent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.