Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,1′,1′′,1′′′-[1,4-Piperazinediylbis(2,1-ethanediylnitrilo)]tetrakis[2-dodecanol] | 1265904-26-4 | 98.0% | 909.54 | 25 MG
SDP

Catalog No. 5000439070
Encompass_Preferred

1,1′,1′′,1′′′-[1,4-Piperazinediylbis(2,1-ethanediylnitrilo)]tetrakis[2-dodecanol] is a lipid/lipidoid used in preparation of lipid-based or lipidoid nanoparticles.

  • Purity of 98.0%
  • Molecular weight of 909.54
  • Appears as a solid
  • Color is light yellow to orange
  • Ships at room temperature in continental US
  • Storage conditions: powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-39070 Supplier Medchemexpress LLC Supplier No. 25MGHY112773
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,1′,1′′,1′′′-[1,4-Piperazinediylbis(2,1-ethanediylnitrilo)]tetrakis[2-dodecanol] is a lipid/lipidoid used in preparation of lipid-based or lipidoid nanoparticles.

  • Purity of 98.0%
  • Molecular weight of 909.54
  • Appears as a solid
  • Color is light yellow to orange
  • Ships at room temperature in continental US
  • Storage conditions: powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.