Learn More
Medchemexpress LLC 1,1′,1′′,1′′′-[1,4-Piperazinediylbis(2,1-ethanediylnitrilo)]tetrakis[2-dodecanol] | 1265904-26-4 | 98.0% | 909.54 | 5 MG
SDP
Catalog No. 5000364400
Encompass_Preferred
1,1′,1′′,1′′′-[1,4-Piperazinediylbis(2,1-ethanediylnitrilo)]tetrakis[2-dodecanol] is a lipid/lipidoid used in the preparation of lipid-based or lipidoid nanoparticles. It is intended for research use only.
  • Used in the preparation of lipid-based or lipidoid nanoparticles.
  • Suitable for biochemical assay reagents.
  • Applicable in drug delivery.
  • Relevant for metabolic enzyme/protease research.
  • Used in liposome research.
Catalog No. 50-003-64400 Supplier Medchemexpress LLC Supplier No. 5MGHY112773
1,1′,1′′,1′′′-[1,4-Piperazinediylbis(2,1-ethanediylnitrilo)]tetrakis[2-dodecanol] is a lipid/lipidoid used in the preparation of lipid-based or lipidoid nanoparticles. It is intended for research use only.
  • Used in the preparation of lipid-based or lipidoid nanoparticles.
  • Suitable for biochemical assay reagents.
  • Applicable in drug delivery.
  • Relevant for metabolic enzyme/protease research.
  • Used in liposome research.