Learn More
Medchemexpress LLC 1,1′,1′′,1′′′-[1,4-Piperazinediylbis(2,1-ethanediylnitrilo)]tetrakis[2-dodecanol] | 1265904-26-4 | 98.0% | 909.54 | 50 MG
SDP
Catalog No. 5000437403
Encompass_Preferred
1,1′,1′′,1′′′-[1,4-Piperazinediylbis(2,1-ethanediylnitrilo)]tetrakis[2-dodecanol] is a lipid/lipidoid chemical compound primarily used in the preparation of lipid-based or lipidoid nanoparticles. This product is for research use only.
  • Solid appearance, light yellow to orange in color.
  • Suitable for lipid-based or lipidoid nanoparticle preparation.
  • High purity of 98.0%.
  • Soluble in DMSO.
  • Specific storage conditions for powder and solutions to maintain stability.
Catalog No. 50-004-37403 Supplier Medchemexpress LLC Supplier No. HY11277350MG
1,1′,1′′,1′′′-[1,4-Piperazinediylbis(2,1-ethanediylnitrilo)]tetrakis[2-dodecanol] is a lipid/lipidoid chemical compound primarily used in the preparation of lipid-based or lipidoid nanoparticles. This product is for research use only.
  • Solid appearance, light yellow to orange in color.
  • Suitable for lipid-based or lipidoid nanoparticle preparation.
  • High purity of 98.0%.
  • Soluble in DMSO.
  • Specific storage conditions for powder and solutions to maintain stability.