Learn More
Cayman Chemical 1,1,1,1-Kestohexose, Purity: ≥98%, CAS: 62512-19-0, 10 mg
SDP
Catalog No. 502909702
Encompass_Preferred
A fructooligosaccharide with neuroprotective and antidepressant-like activities; prevents corticosterone-induced decreases in viability and NGF expression in PC12 cells at 0.625 and 1.25 µM; reduces immobility time in the forced swim test in rats at 20 mg/kg
Catalog No. 50-290-9702 Supplier Cayman Chemical Supplier No. 3659110 MG
A fructooligosaccharide with neuroprotective and antidepressant-like activities; prevents corticosterone-induced decreases in viability and NGF expression in PC12 cells at 0.625 and 1.25 µM; reduces immobility time in the forced swim test in rats at 20 mg/kg