Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Henicosafluoro-12-Iodododecane

Catalog No. 9198725GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-987-25GF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 91-987-25GF Supplier Sigma Aldrich Supplier No. 919872.5GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Building block to introduce a fluorinated pony tail in compounds for fluorous phase -+immobilization-+ by fluorous phase partition

Specifications

CAS 7146-15-8
Molecular Formula C6H13NO2 -+ HCl
Linear Formula (CH3)2CHCH(NH2)COOCH3 -+ HCl
MDL Number MFCD00237309
Quantity 50 mg
Synonym 1-Iodo-1H,1H,2H,2H-perfluorododecane; 1H,1H,2H,2H-Perfluorododecyl iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.