Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,2,3,3,3-Heptachloropropane 98.0+%, TCI America™
SDP

Catalog No. H001610G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H001610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H001610G Supplier TCI America Supplier No. H001610G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3849-33-0
Molecular Formula C3HCl7
Molecular Weight (g/mol) 285.19
MDL Number MFCD00058935
InChI Key LZOUTMTXULWVSU-UHFFFAOYSA-N
PubChem CID 77457
IUPAC Name 1,1,1,2,3,3,3-heptachloropropane
SMILES ClC(C(Cl)(Cl)Cl)C(Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 249°C
Quantity 10 g
Formula Weight 285.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,2,3,3,3-Heptachloropropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.