Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,16,16,17,17,18,18,18-Tricosafluoro-5,8,11,14-tetrakis(trifluoromethyl)-3,6,9,12,15 95+%, TCI America™
SDP

Catalog No. T353825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T353825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T353825G Supplier TCI America Supplier No. T353825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 37486-69-4
Molecular Formula C17HF35O5
Molecular Weight (g/mol) 950.13
MDL Number MFCD00054716
InChI Key MEXQRXXROOSHGK-UHFFFAOYNA-N
Synonym 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,16,16,17,17,18,18,18-tricosafluoro-5,8,11,14-tetrakis trifluoromethyl-3,6,9,12,15-pentaoxaoctadecane, 2h-perfluoro-5,8,11,14-tetramethyl-3,6,9,12,15-pentaoxaoctadecane, 3,6,9,12,15-pentaoxaoctadecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,16,16,17,17,18,18,18-tricosafluoro-5,8,11,14-tetrakis trifluoromethyl, 1,1,1,2,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy propoxy-3-1,2,2,2-tetrafluoroethoxy propane, unii-sef36f005e, freon e-5, 2h-tricosafluoro-5,8,11,14-tetrakis trifluoromethyl-3,6,9,12,15-pentaoxaoctadecane, 1,1,1,2,3,3-hexakis fluoranyl-2-1,1,2,3,3,3-hexakis fluoranyl-2-1,1,2,3,3,3-hexakis fluoranyl-2-1,1,2,3,3,3-hexakis fluoranyl-2-1,1,2,2,3,3,3-heptakis fluoranyl propoxy propoxy propoxy propoxy-3-1,2,2,2-tetrakis fluoranyl ethoxy propane
PubChem CID 123459
IUPAC Name 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,16,16,17,17,18,18,18-tricosafluoro-5,8,11,14-tetrakis(trifluoromethyl)-3,6,9,12,15-pentaoxaoctadecane
SMILES FC(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

Color Colorless
Boiling Point 224°C
Quantity 25 g
Formula Weight 950.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,16,16,17,17,18,18,18-Tricosafluoro-5,8,11,14-tetrakis(trifluor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.