Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol 98.0+%, TCI America™
SDP

Catalog No. H134925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H134925G 25 g
H13495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H134925G Supplier TCI America Supplier No. H134925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1515-14-6
Molecular Formula C4H4F6O
Molecular Weight (g/mol) 182.07
MDL Number MFCD00039259
InChI Key FQDXJYBXPOMIBX-UHFFFAOYSA-N
Synonym hexafluoro-2-methylisopropanol, 1,1,1,3,3,3-hexafluoro-2-methyl-2-propanol, 2-propanol, 1,1,1,3,3,3-hexafluoro-2-methyl, hexafluoro-tert-butanol, cf3 2mecoh, 1,1,1,3,3,3-hexafluoro-2-methylisopropanol, methylbistrifluoromethylmethanol, cf3 2c oh ch3, fqdxjybxpomibx-uhfffaoysa, hexafluoro-3-methylisopropanol
PubChem CID 73936
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-methylpropan-2-ol
SMILES CC(O)(C(F)(F)F)C(F)(F)F

Specifications

Color Colorless
Boiling Point 62°C
Quantity 25 g
Formula Weight 182.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.