Learn More
Medchemexpress LLC (1,1,1,3,3,3-hexafluoro-2-methylpropan-2-yl) 4-[(4-hydroxy-2-oxochromen-3-yl)methyl]benzoate | 867257-26-9 | MFCD16620516 | 98.7% | 460.32 g/mol | C21H14F6O5 | 10 MG
SDP
Catalog No. 5000286637
Encompass_Preferred
Tecarfarin is an analytical research standard of a non-competitive vitamin K epoxide reductase (VKOR) antagonist used in anticoagulant research. Supplied as a powder, the product is intended for in vitro studies, assay development, and use as a reference material. The 10 mg pack can be reconstituted to a 1 mM solution in DMSO (≈21.7240 mL); storage recommendations are provided for both powder and solutions.
Catalog No. 50-002-86637 Supplier Medchemexpress LLC Supplier No. HY1485410MG
Tecarfarin is an analytical research standard of a non-competitive vitamin K epoxide reductase (VKOR) antagonist used in anticoagulant research. Supplied as a powder, the product is intended for in vitro studies, assay development, and use as a reference material. The 10 mg pack can be reconstituted to a 1 mM solution in DMSO (≈21.7240 mL); storage recommendations are provided for both powder and solutions.