Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,3,3,3-Hexafluoroisopropanol, >99%, MP Biomedicals™

Catalog No. ICN15124580 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15124580 100 mL
ICN15124505 5 mL
ICN15124525 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15124580 Supplier MP Biomedicals, Inc Supplier No. 0215124580
Add to Cart
Add to Cart

1,1,1,3,3,3-Hexafluoro-Isopropanol is a speciality solvent effective for solubilizing a wide range of polymers, including those that are not soluble in the most common organic solvents.

  • 1,1,1,3,3,3-Hexafluoro-Isopropanol is a speciality solvent effective for solubilizing a wide range of polymers, including those that are not soluble in the most common organic solvents, such as:
    • Polyamides, polyacrylonitriles, polyacetals, polyesters (e.g. polyglycolide), and polyketones.
  • It has also found use in biochemistry to solubilize peptides and to monomerize β-sheet protein aggregates.
  • It can be used as an acid in volatile buffers for ion pair HPLC - mass spectrometry of nucleic acids.
  • 1,1,1,3,3,3-Hexafluoro-2-propanol was used for preparing hexafluoroalcohol-functionalized methacrylate polymers for lithographic/nanopatterning materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29463-77-2
Molecular Formula C3H2F6O
Molecular Weight (g/mol) 168.038
InChI Key BYEAHWXPCBROCE-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoro-2-propanol, hexafluoroisopropanol, hfip, hexafluoro-2-propanol, hexafluoroisopropyl alcohol, bis trifluoromethyl methanol, 1,1,1,3,3,3-hexafluoroisopropanol, 2h-hexafluoroisopropanol, 2-propanol, 1,1,1,3,3,3-hexafluoro, 1,1,1,3,3,3-hexafluoroisopropyl alcohol
PubChem CID 13529
ChEBI CHEBI:63104
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
SMILES C(C(F)(F)F)(C(F)(F)F)O

Specifications

Color Colorless
Density 1.62 g/mL
Assay Percent Range ≥99%
Quantity 100 mL
Formula Weight 168.04
Percent Purity 0.9969
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-isopropanol, For GC analysis
Recommended Storage Store at room temperature (15°C to 30°C).

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.