Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,3,3,3-Hexafluoroisopropyl acrylate, 98+%, stab. with 50ppm 4-Methoxyphenol, Thermo Scientific™

Catalog No. AAB2139614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2139614 25 g
AAB2139603 1 g
AAB2139606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2139614 Supplier Thermo Scientific Chemicals Supplier No. B2139614

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2160-89-6
Molecular Formula C6H4F6O2
Molecular Weight (g/mol) 222.09
MDL Number MFCD00040104
InChI Key MNSWITGNWZSAMC-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoroisopropyl acrylate, hexafluoroisopropyl acrylate, 1,1,1,3,3,3-hexafluoropropan-2-yl acrylate, 2-propenoic acid, 2,2,2-trifluoro-1-trifluoromethyl ethyl ester, acrylic acid 1,1,1,3,3,3-hexafluoroisopropyl ester, hexafluoro-2-propyl acrylate, mnswitgnwzsamc-uhfffaoysa, 1h-1-trifluoromethyl trifluoroethyl acrylate
PubChem CID 75096
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-yl prop-2-enoate
SMILES FC(F)(F)C(OC(=O)C=C)C(F)(F)F

Specifications

Boiling Point 73°C to 75°C
Flash Point 10°C (50°F)
Linear Formula CH2=CHCO2CH(CF3)2
Quantity 25 g
UN Number UN3272
Beilstein 2094742
Formula Weight 222.09
Percent Purity ≥98%
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropyl acrylate, stabilized with 50ppm 4-Methoxyphenol
Hazard Category Danger
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.