Learn More
Medchemexpress LLC 1,1,1,3,3,3-hexafluoropropan-2-yl 4-(3-phenoxybenzyl)piperazine-1-carboxylate | 1416133-89-5 | MFCD22987956 | 99.9% | 462.39 | C21H20F6N2O3 | 50MG
SDP
Catalog No. 5000239030
Encompass_Preferred
JW-642 is a research-grade, selective inhibitor of monoacylglycerol lipase (MAGL) used to probe endocannabinoid signaling and lipid metabolism. It provides low-nanomolar potency against MAGL across species while showing much weaker activity against fatty acid amide hydrolase, supporting its use as a selective pharmacological tool.
  • Potent MAGL inhibition in human, mouse, and rat (IC50: 3.7 nM human; 7.6 nM mouse; 14 nM rat).
  • Reduced activity versus FAAH (micromolar IC50s), indicating selectivity.
  • High purity (99.9% by HPLC) for consistent experimental performance.
  • Solid, stable material suitable for analytical and preparative applications.
  • Recommended storage under nitrogen at 4°C; in solution store at -80°C for long-term stability.
  • Suitable for biochemical, enzymatic, and cell-based assays.
Catalog No. 50-002-39030 Supplier Medchemexpress LLC Supplier No. HY1233250MG
JW-642 is a research-grade, selective inhibitor of monoacylglycerol lipase (MAGL) used to probe endocannabinoid signaling and lipid metabolism. It provides low-nanomolar potency against MAGL across species while showing much weaker activity against fatty acid amide hydrolase, supporting its use as a selective pharmacological tool.
  • Potent MAGL inhibition in human, mouse, and rat (IC50: 3.7 nM human; 7.6 nM mouse; 14 nM rat).
  • Reduced activity versus FAAH (micromolar IC50s), indicating selectivity.
  • High purity (99.9% by HPLC) for consistent experimental performance.
  • Solid, stable material suitable for analytical and preparative applications.
  • Recommended storage under nitrogen at 4°C; in solution store at -80°C for long-term stability.
  • Suitable for biochemical, enzymatic, and cell-based assays.