Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,1,1,3,3,3-hexafluoropropan-2-yl 4-(3-phenoxybenzyl)piperazine-1-carboxylate | 1416133-89-5 | MFCD22987956 | 99.9% | 462.39 g·mol⁻¹ | C21H20F6N2O3 | 100MG
SDP

Catalog No. 5000237097
Encompass_Preferred

JW 642 is a potent inhibitor of monoacylglycerol lipase (MAGL) provided as a research-grade analytical standard for in vitro and analytical applications. It is intended for laboratory research use only and not for human or clinical use. Recommended handling includes storage under inert atmosphere and preparation in anhydrous solvent to maintain stability.

  • Potent MAGL inhibitor with low-nanomolar activity.
  • High purity: 99.9%.
  • Molecular weight: 462.39 g·mol⁻¹.
  • Chemical formula: C21H20F6N2O3.
  • Supplied as a 100 mg research standard.
  • Soluble in DMSO at ≥ 100 mg/mL; hygroscopic DMSO may affect solubility.
  • Store solid under nitrogen at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-37097 Supplier Medchemexpress LLC Supplier No. HY12332100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JW 642 is a potent inhibitor of monoacylglycerol lipase (MAGL) provided as a research-grade analytical standard for in vitro and analytical applications. It is intended for laboratory research use only and not for human or clinical use. Recommended handling includes storage under inert atmosphere and preparation in anhydrous solvent to maintain stability.

  • Potent MAGL inhibitor with low-nanomolar activity.
  • High purity: 99.9%.
  • Molecular weight: 462.39 g·mol⁻¹.
  • Chemical formula: C21H20F6N2O3.
  • Supplied as a 100 mg research standard.
  • Soluble in DMSO at ≥ 100 mg/mL; hygroscopic DMSO may affect solubility.
  • Store solid under nitrogen at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.