Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,1,1,3,3,3-hexafluoropropan-2-yl 4-[(3-phenoxyphenyl)methyl]piperazine-1-carboxylate | 1416133-89-5 | MFCD22987956 | 99.9% | 462.39 g/mol | C21H20F6N2O3 | 25MG
SDP

Catalog No. 5000235485
Encompass_Preferred

JW 642 is a potent, selective inhibitor of monoacylglycerol lipase (MAGL) intended for research use. It is supplied as a high-purity solid and is used in biochemical and pharmacological studies to probe MAGL activity in vitro and in vivo.

  • Potent MAGL inhibitor; IC50: mouse 7.6 nM, rat 14 nM, human 3.7 nM.
  • Selective over FAAH; FAAH IC50s: mouse 31 μM, rat 14 μM, human 20.6 μM.
  • Purity 99.90%.
  • Molecular formula C21H20F6N2O3; molecular weight 462.39 g/mol.
  • Appearance white to off-white solid.
  • Solubility DMSO ≥100 mg/mL; in vivo formulations achieve ≥2.5 mg/mL.
  • For research use only.

Catalog No. 50-002-35485 Supplier Medchemexpress LLC Supplier No. HY1233225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

JW 642 is a potent, selective inhibitor of monoacylglycerol lipase (MAGL) intended for research use. It is supplied as a high-purity solid and is used in biochemical and pharmacological studies to probe MAGL activity in vitro and in vivo.

  • Potent MAGL inhibitor; IC50: mouse 7.6 nM, rat 14 nM, human 3.7 nM.
  • Selective over FAAH; FAAH IC50s: mouse 31 μM, rat 14 μM, human 20.6 μM.
  • Purity 99.90%.
  • Molecular formula C21H20F6N2O3; molecular weight 462.39 g/mol.
  • Appearance white to off-white solid.
  • Solubility DMSO ≥100 mg/mL; in vivo formulations achieve ≥2.5 mg/mL.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.