Learn More
Medchemexpress LLC 1,1,1,3,3,3-hexafluoropropan-2-yl 4-[(3-phenoxyphenyl)methyl]piperazine-1-carboxylate | 1416133-89-5 | MFCD22987956 | 99.9% | 462.39 g/mol | C21H20F6N2O3 | 5MG
SDP
Catalog No. 5000239185
Encompass_Preferred
JW 642 is a small-molecule monoacylglycerol lipase (MAGL) inhibitor provided as a white to off-white solid for laboratory research. It has chemical formula C21H20F6N2O3 and molecular weight 462.39 g/mol, with reported purity of 99.9%. The compound is highly soluble in DMSO and can be formulated for in vivo dosing using common vehicles. Store under nitrogen at 4°C; prepared solutions are stable for extended storage at -80°C.
Catalog No. 50-002-39185 Supplier Medchemexpress LLC Supplier No. HY123325MG
JW 642 is a small-molecule monoacylglycerol lipase (MAGL) inhibitor provided as a white to off-white solid for laboratory research. It has chemical formula C21H20F6N2O3 and molecular weight 462.39 g/mol, with reported purity of 99.9%. The compound is highly soluble in DMSO and can be formulated for in vivo dosing using common vehicles. Store under nitrogen at 4°C; prepared solutions are stable for extended storage at -80°C.