Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,3,3,3-Hexamethyldisilazane, HMDS, EMD-Millipore

Catalog No. M1121860025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1121860025 25 mL Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1121860025 Supplier MilliporeSigma Supplier No. 1.12186.0025

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 999-97-3
Molecular Formula C6H19NSi2
Molecular Weight (g/mol) 161.395
InChI Key FFUAGWLWBBFQJT-UHFFFAOYSA-N
Synonym Bis(trimethylsilyl)amine, HMDS
PubChem CID 13838
ChEBI CHEBI:85068
IUPAC Name [dimethyl-(trimethylsilylamino)silyl]methane
SMILES C[Si](C)(C)N[Si](C)(C)C

Specifications

Packaging Glass
Explosion Limit 0.8 to 25.9 %(V)
Ignition Point 325°C
Vapor Pressure 20 hPa (20°C)
Viscosity 0.9mm2/sec., kinematic (25°C)
Density Density: 0.774g/cm3 (20°C)
Quantity 25 mL
Formula Weight 161.39
Chemical Name or Material 1,1,1,3,3,3-Hexamethyldisilazane
Recommended Storage Store at 15° to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.