Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,1,1,3,5,5,5-Heptamethyltrisiloxane

Catalog No. 37088650ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-088-650ML 50 mL
37-088-6250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 37-088-650ML Supplier Sigma Aldrich Supplier No. 37088650ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,1,1,3,5,5,5-Heptamethyltrisiloxane can be used for the aromatic C-H silylation of arenes in the presence of a platinum complex catalyst.It can also be used as a reagent to synthesize: 3-Octyl-1,1,1,3,5,5,5-heptamethyltrisiloxane, an agricultural adjuvant and sensory performance enhancer used in cosmetic formulations.Isoindigo-based conjugated polymer with siloxane-terminated solubilizing group.Monosiloxy esters with a variety of acids.

Specifications

CAS 1873-88-7
Density 0.819 g/mL at 25°C
Boiling Point 142°C
Molecular Formula C7H22O2Si3
Refractive Index n20/D 1.382
Linear Formula [(CH3)3SiO]2SiHCH3
MDL Number MFCD00048000
Quantity 50 mL
Molecular Weight (g/mol) 222.5 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.