Learn More
Medchemexpress LLC 1,1'-(1,4-Phenylene)bis(3-(1H-indol-4-yl)urea) | 3095291-54-3 | 98.0% | 424.45 | 25 MG
SDP
Catalog No. 5000394815
Encompass_Preferred
α-Synuclein inhibitor 11 (compound 1) is a selective α-synuclein (α-syn) oligomer formation inhibitor. It does not inhibit tau 4R (isoforms 0N4R, 2N4R) or p-tau (isoform 1N4R), making it a valuable tool for Parkinson's disease (PD) research. This product is intended for research use only.
  • Selective α-synuclein oligomer formation inhibitor
  • Does not inhibit tau 4R or p-tau
  • Suitable for Parkinson's disease research
Catalog No. 50-003-94815 Supplier Medchemexpress LLC Supplier No. 25MGHY163145
Add to Cart
Add to Cart
α-Synuclein inhibitor 11 (compound 1) is a selective α-synuclein (α-syn) oligomer formation inhibitor. It does not inhibit tau 4R (isoforms 0N4R, 2N4R) or p-tau (isoform 1N4R), making it a valuable tool for Parkinson's disease (PD) research. This product is intended for research use only.
  • Selective α-synuclein oligomer formation inhibitor
  • Does not inhibit tau 4R or p-tau
  • Suitable for Parkinson's disease research