Learn More
Medchemexpress LLC 1,1'-(1,4-Phenylene)bis(3-(1H-indol-4-yl)urea) | 3095291-54-3 | 98.0% | 424.45 | 50 MG
SDP
Catalog No. 5000394503
Encompass_Preferred
α-Synuclein inhibitor 11 (compound 1) is a selective α-synuclein (α-syn) oligomer formation inhibitor. It does not inhibit tau 4R (isoforms 0N4R, 2N4R) or p-tau (isoform 1N4R). This product can be used for Parkinson's disease (PD) research.
  • Selective α-synuclein (α-syn) oligomer formation inhibitor.
  • Does not inhibit tau 4R (isoforms 0N4R, 2N4R) or p-tau (isoform 1N4R).
  • Applicable for Parkinson's disease (PD) research.
Catalog No. 50-003-94503 Supplier Medchemexpress LLC Supplier No. HY16314550MG
α-Synuclein inhibitor 11 (compound 1) is a selective α-synuclein (α-syn) oligomer formation inhibitor. It does not inhibit tau 4R (isoforms 0N4R, 2N4R) or p-tau (isoform 1N4R). This product can be used for Parkinson's disease (PD) research.
  • Selective α-synuclein (α-syn) oligomer formation inhibitor.
  • Does not inhibit tau 4R (isoforms 0N4R, 2N4R) or p-tau (isoform 1N4R).
  • Applicable for Parkinson's disease (PD) research.