Learn More
Medchemexpress LLC 1-(1,1-dimethylethyl) tridecanedioate | 2703752-86-5 | 300.43 | C17H32O4 | 500mg
SDP
Catalog No. 5000268372
Encompass_Preferred
1-(1 1-Dimethylethyl) tridecanedioate is a PROTAC Linker and can be used for PROTAC synthesis such as FF2039 (HY-172159)[1]
Catalog No. 50-002-68372 Supplier Medchemexpress LLC Supplier No. HYW881439500MG
1-(1 1-Dimethylethyl) tridecanedioate is a PROTAC Linker and can be used for PROTAC synthesis such as FF2039 (HY-172159)[1]