Learn More
Medchemexpress LLC 1-(1,1-dimethylethyl) tridecanedioate | 2703752-86-5 | MFCD30737823 | 98.0% | 300.43 | C17H32O4 | 5g
SDP
Catalog No. 5000270359
Encompass_Preferred
1-(1 1-Dimethylethyl) tridecanedioate is a PROTAC Linker and can be used for PROTAC synthesis such as FF2039 (HY-172159)[1]
Catalog No. 50-002-70359 Supplier Medchemexpress LLC Supplier No. HYW8814395G
1-(1 1-Dimethylethyl) tridecanedioate is a PROTAC Linker and can be used for PROTAC synthesis such as FF2039 (HY-172159)[1]