Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,1,1-Trichloro-2-methyl-2-propanol hemihydrate

Catalog No. 112054250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-205-4250G 250 g
11-205-4500G 500 g
11-205-450G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-205-4250G Supplier Sigma Aldrich Supplier No. 112054250G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate

Specifications

CAS 6001-64-5
Melting Point 77°C to 79°C (lit.)
Molecular Formula C4H7Cl3O -+ 0.5H2O
Linear Formula Cl3CC(CH3)2 OH -+ 0.5H2O
MDL Number MFCD02179352
Quantity 250 g
Synonym beta,beta,beta-Trichloro-t-butanol; beta,beta,beta-Trichloro-tert-butyl alcohol hemihydrate; Acetone chloroform; Chloretone; Chlorobutanol
Molecular Weight (g/mol) 186.46
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.