Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1-Trichloroethane in Dimethyl Sulfoxide, SPEX CertiPrep™
SDP

Catalog No. USPS3605DMS Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USPS3605DMS 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USPS3605DMS Supplier SPEX CertiPrep Supplier No. USPS3605DMSO

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,1,1-Trichloroethane71-55-650,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 71-55-6
Molecular Formula C2H3Cl3
Molecular Weight (g/mol) 133.396
InChI Key UOCLXMDMGBRAIB-UHFFFAOYSA-N
PubChem CID 6278
ChEBI CHEBI:36015
IUPAC Name 1,1,1-trichloroethane
SMILES CC(Cl)(Cl)Cl

Specifications

Packaging 2mL Amber Prescored Glass Ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Explosion Limit 1.8% (volume); 63% (volume)
Ignition Point 270°C
Vapor Pressure 2.5 hPA (20°C)
Analyte or Component Names 1,1,1-Trichloroethane
Density 1.1g/cm3
Physical Form Fluid
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Element 1,1,1-Trichloroethane
Solvent or Matrix Dimethyl Sulfoxide
Quantity 1 mL
Grade USP
Concentration or Composition (by Analyte or Components) 50, 000μg/mL
Chemical Name or Material 1,1,1-Trichloroethane in Dimethyl Sulfoxide, USP 467 Standard
Show More Show Less
DOTInformation Nonhazardous
Recommended Storage Freezer

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.