Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,1,1-Trifluoro-2,4-pentanedione

Catalog No. 23597025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-597-025G 25 g
23-597-010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-597-025G Supplier Sigma Aldrich Supplier No. 23597025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

May darken in storage1,1,1-Trifluoro-2,4-pentanedione has been used as reagent in the preparation of 2-alkylcarbonyl and 2-benzoyl-3-trifluoromethylquinoxaline 1,4-di-N-oxide derivatives.

Specifications

CAS 367-57-7
Density 1.27 g/mL (at 25°C (literature))
Boiling Point 105°C to 107°C (lit.)
Molecular Formula C5H5F3O2
Refractive Index n20/D 1.388 (literature)
Linear Formula CH3COCH2COCF3
MDL Number MFCD00000427
Quantity 25 g
Synonym alpha,alpha,alpha-Trifluoroacetylacetone; 1,1,1-Trifluoroacetylacetone
Molecular Weight (g/mol) 154.09
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.