Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1-Trifluoro-2-propanol, 97%

Catalog No. aal1115006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1115006 5 g
AAL1115014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1115006 Supplier Thermo Scientific Chemicals Supplier No. L1115006
Only null left
Add to Cart

CAS: 374-01-6 | C3H5F3O | 114.067 g/mol

1,1,1-Trifluoro-2-propanol is used as a chemical building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1-Trifluoro-2-propanol is used as a chemical building block.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374-01-6
Molecular Formula C3H5F3O
Molecular Weight (g/mol) 114.067
MDL Number MFCD00014403
InChI Key GILIYJDBJZWGBG-UHFFFAOYSA-N
Synonym 1,1,1-trifluoro-2-propanol, 2-propanol, 1,1,1-trifluoro, 1,1,1-trifluoroisopropanol, 1,1,1-trifluoropropanol-2, 1,1,1-trifluoro-propan-2-ol, 1-methyl-2,2,2-trifluoroethanol, pubchem12648, acmc-20ap8u, 1,1-trifluoro-2-propanol, 2-propanol,1,1-trifluoro
PubChem CID 9774
IUPAC Name 1,1,1-trifluoropropan-2-ol
SMILES CC(C(F)(F)F)O

Specifications

Density 1.259
Boiling Point 75°C to 76°C
Flash Point 25°C (77°F)
Refractive Index 1.3175
Quantity 5 g
UN Number UN1987
Beilstein 1734738
Solubility Information Fully miscible in water.
Formula Weight 114.07
Percent Purity 97%
Chemical Name or Material 1,1,1-Trifluoro-2-propanol
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 206-773-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.