Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,1,1-Trifluoroacetone

Catalog No. T628045G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T628045G 5 g
T62804100G 100 g
T6280425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T628045G Supplier Sigma Aldrich Supplier No. T628045G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Used in a synthesis of 2-trifluoromethyl-7-azaindoles starting with 2,6-dihalopyridines
  • The derived chiral imine was used to prepare enantiopure α-trifluoromethyl alanines and diamines via a strecker reaction followed by either nitrile hydrolysis or reduction

Specifications

CAS 421-50-1
Density 1.252 g/mL (at 25°C (literature))
Boiling Point 22°C (lit.)
Molecular Formula C3H3F3O
Refractive Index n20/D 1.3 (literature)
Linear Formula CH3COCF3
MDL Number MFCD00000423
Quantity 5 g
Molecular Weight (g/mol) 112.05
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.