Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1-Trifluoroacetone, 95%

Catalog No. AAA1355622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1355622 100 g
AAA1355614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1355622 Supplier Thermo Scientific Chemicals Supplier No. A1355622
Add to Cart

CAS: 421-50-1 | C3H3F3O | 112.051 g/mol

1,1,1-Trifluoroacetone is used in a synthesis of 2-trifluoromethyl-7-azaindoles starting with 2,6-dihalopyridines. The derived chiral imine was used to prepare enantiopure α-trifluoromethyl alanines and diamines via a Strecker reaction followed by either nitrile hydrolysis or reduction. And also used as a useful molecule in synthesis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1-Trifluoroacetone is used in a synthesis of 2-trifluoromethyl-7-azaindoles starting with 2,6-dihalopyridines. The derived chiral imine was used to prepare enantiopure α-trifluoromethyl alanines and diamines via a Strecker reaction followed by either nitrile hydrolysis or reduction. And also used as a useful molecule in synthesis reactions.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 421-50-1
Molecular Formula C3H3F3O
Molecular Weight (g/mol) 112.051
MDL Number MFCD00000423
InChI Key FHUDAMLDXFJHJE-UHFFFAOYSA-N
Synonym 1,1,1-trifluoroacetone, trifluoroacetone, 1,1,1-trifluoro-2-propanone, methyl trifluoromethyl ketone, 2-propanone, 1,1,1-trifluoro, trifluoromethyl methyl ketone, 3,3,3-trifluoroacetone, 1,1,1,-trifluoroacetone, trifluoroketone, trifluoracetone
PubChem CID 9871
IUPAC Name 1,1,1-trifluoropropan-2-one
SMILES CC(=O)C(F)(F)F

Specifications

Melting Point <-78°C
Density 1.252
Boiling Point 21°C to 24°C
Flash Point −31°C (−24°F)
Odor Chloroform-like
Refractive Index 1.3
Quantity 100 g
UN Number UN1993
Beilstein 1748614
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 112.05
Percent Purity 95%
Chemical Name or Material 1,1,1-Trifluoroacetone
Show More Show Less
Hazard Category H224-H314-H335
Hazard Statement GHS H Statement
H224-H315-H319-H335
Extremely flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 207-005-9
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.