Learn More
Strem, An Ascensus Company CAS# 367-57-7. 10g. 1,1,1-Trifluoroacetylacetone, min. 98%. MFCD00000427
SDP
Catalog No. 5090109427
Encompass
CAS# 367-57-7. 10g. 1,1,1-Trifluoroacetylacetone, min. 98%. MFCD00000427. Molecular Weight: 154.09. Molecular Formula: CF3COCH2COCH3. Color/form: colorless liq. Strem# 09-7210. http://www.strem.com/catalog/v/09-7210/
Catalog No. 50-901-09427 Supplier Strem, An Ascensus Company Supplier No. 09721010G
CAS# 367-57-7. 10g. 1,1,1-Trifluoroacetylacetone, min. 98%. MFCD00000427. Molecular Weight: 154.09. Molecular Formula: CF3COCH2COCH3. Color/form: colorless liq. Strem# 09-7210. http://www.strem.com/catalog/v/09-7210/