Learn More
Medchemexpress LLC 1,1,1-Trifluoroethyl-PEG2-azide | 1835759-68-6 | 97.0% | C6H10F3N3O2 | 50 MG
SDP
Catalog No. 5000355403
Encompass_Preferred
1,1,1-Trifluoroethyl-PEG2-azide is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs and functions as a click chemistry reagent.
  • Functions as a click chemistry reagent
  • Contains an azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
  • Used in the synthesis of PROTACs
  • PROTACs degrade specific proteins via the ubiquitin-proteasome system
Catalog No. 50-003-55403 Supplier Medchemexpress LLC Supplier No. HY14084550MG
1,1,1-Trifluoroethyl-PEG2-azide is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs and functions as a click chemistry reagent.
  • Functions as a click chemistry reagent
  • Contains an azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
  • Used in the synthesis of PROTACs
  • PROTACs degrade specific proteins via the ubiquitin-proteasome system