Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,1':2',1''-terphenyl-d14 | 5142-67-6 | 99% | 244.39 | 10 MG
SDP

Catalog No. 5000366913
Encompass_Preferred

1,1':2',1''-Terphenyl-d14 is the deuterium labeled 1,1':2',1''-Terphenyl.

  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs

Catalog No. 50-003-66913 Supplier Medchemexpress LLC Supplier No. HYW009658S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,1':2',1''-Terphenyl-d14 is the deuterium labeled 1,1':2',1''-Terphenyl.

  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.