Learn More
Medchemexpress LLC 1,1':2',1''-Terphenyl-d14 | 5142-67-6 | 99% | 244.39 | 100 MG
SDP
Catalog No. 5000451547
Encompass_Preferred
1,1':2',1''-Terphenyl-d14 is the deuterium labeled 1,1':2',1''-Terphenyl. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-51547 Supplier Medchemexpress LLC Supplier No. HYW009658S100MG
1,1':2',1''-Terphenyl-d14 is the deuterium labeled 1,1':2',1''-Terphenyl. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.