Learn More
Medchemexpress LLC 1,1':2',1''-Terphenyl-d14 | 5142-67-6 | 99% | 244.39 | 25 MG
SDP
Catalog No. 5000454017
Encompass_Preferred
1,1':2',1''-Terphenyl-d14 is a deuterium-labeled compound, specifically a stable isotope of 1,1':2',1''-Terphenyl. It is intended for research use only and not for medical applications. This compound is valuable as a tracer and an internal standard in various quantitative analyses, including NMR, GC-MS, and LC-MS. The incorporation of stable heavy isotopes like deuterium into molecules can influence their pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Acts as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeling can impact pharmacokinetic and metabolic properties.
  • For research use only.
Catalog No. 50-004-54017 Supplier Medchemexpress LLC Supplier No. 25MGHYW009658S
1,1':2',1''-Terphenyl-d14 is a deuterium-labeled compound, specifically a stable isotope of 1,1':2',1''-Terphenyl. It is intended for research use only and not for medical applications. This compound is valuable as a tracer and an internal standard in various quantitative analyses, including NMR, GC-MS, and LC-MS. The incorporation of stable heavy isotopes like deuterium into molecules can influence their pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Acts as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeling can impact pharmacokinetic and metabolic properties.
  • For research use only.