Learn More
Medchemexpress LLC 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane-1-sulfonamide | 41997-13-1 | MFCD09263736 | >75.0% | 399.13 g·mol⁻¹ | C6H2F13NO2S | 5 MG
SDP
Catalog No. 5000283968
Encompass_Preferred
Perfluorohexane sulfonamide is a perfluoroalkyl sulfonamide supplied as a powder for laboratory research use. It is used in analytical and biological studies of PFAS and has been reported to inhibit carbonic anhydrase. Handle as a research chemical and follow the safety data sheet for storage, handling, and disposal instructions.
  • Perfluoroalkyl sulfonamide research reagent for analytical and biological studies.
  • Reported inhibitor of carbonic anhydrase suitable for in vitro experiments.
  • Powder form with recommended storage at -20 °C; store solutions at -80 °C.
  • Chemical formula C6H2F13NO2S; molecular weight 399.13 g·mol⁻¹.
  • Not for human or clinical use; follow SDS for handling and disposal.
Catalog No. 50-002-83968 Supplier Medchemexpress LLC Supplier No. HYW5878055MG
Perfluorohexane sulfonamide is a perfluoroalkyl sulfonamide supplied as a powder for laboratory research use. It is used in analytical and biological studies of PFAS and has been reported to inhibit carbonic anhydrase. Handle as a research chemical and follow the safety data sheet for storage, handling, and disposal instructions.
  • Perfluoroalkyl sulfonamide research reagent for analytical and biological studies.
  • Reported inhibitor of carbonic anhydrase suitable for in vitro experiments.
  • Powder form with recommended storage at -20 °C; store solutions at -80 °C.
  • Chemical formula C6H2F13NO2S; molecular weight 399.13 g·mol⁻¹.
  • Not for human or clinical use; follow SDS for handling and disposal.