Learn More
Medchemexpress LLC 1,1'-(2,2-Dichloroethene-1,1-diyl)bis(4-chlorobenzene)-2,3,5,6-d4 | 93952-19-3 | 99.0% | 1 MG
SDP
Catalog No. 5000411819
Encompass_Preferred
p,p'-DDE-d8 is a deuterium-labeled version of p,p'-DDE, which is a significant metabolite of dichlorodiphenyltrichloroethane (DDT). This compound acts as a potent antagonist for the androgen receptor. It can be utilized as a tracer in research and as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for quantitation during drug development.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11819 Supplier Medchemexpress LLC Supplier No. HYB1986S1MG
p,p'-DDE-d8 is a deuterium-labeled version of p,p'-DDE, which is a significant metabolite of dichlorodiphenyltrichloroethane (DDT). This compound acts as a potent antagonist for the androgen receptor. It can be utilized as a tracer in research and as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for quantitation during drug development.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.