Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2-Tetrabromoethane (Laboratory), Fisher Chemical™

Catalog No. A33500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A33-500 500 mL Glass Bottle
A33-4 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. A33-500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-27-6
Molecular Formula C2H2Br4
Molecular Weight (g/mol) 345.65
MDL Number MFCD00000133
InChI Key QXSZNDIIPUOQMB-UHFFFAOYSA-N
Synonym acetylene tetrabromide, muthmann's liquid, ethane, 1,1,2,2-tetrabromo, tetrabromoacetylene, sym-tetrabromoethane, s-tetrabromoethane, 1,1,2,2-tetrabromaethan, 1,1,2,2-tetrabromoetano, tbe, unii-v00gkc584o
PubChem CID 6588
IUPAC Name 1,1,2,2-tetrabromoethane
SMILES BrC(Br)C(Br)Br

Specifications

Alpha Vector Tetrabromoethane
Melting Point 1°C
Color Yellow
Density 2.960g/cm³
Boiling Point 244°C
Packaging Glass Bottle
Linear Formula CHBr2CHBr2
Quantity 500 mL
UN Number 2504
Formula Weight 345.67
Grade Laboratory
Physical Form Liquid
Chemical Name or Material 1,1,2,2-Tetrabromoethane
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 6.1, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.