Learn More
Cambridge Isotope Laboratories 1,1,2,2-Tetrachloroethane-D2, 33685-54-0, MFCD00037672, 10 g
Catalog No. NC9061958
Encompass_Preferred
Chemical Formula: Cl2CDCDCl2, MW: 169.86, 99.5%.
Catalog No. NC9061958 Supplier Cambridge Isotope Laboratories Supplier No. DLM3510
Chemical Formula: Cl2CDCDCl2, MW: 169.86, 99.5%.