Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2-Tetrachloroethane-d{2}, 99.5% (Isotopic)

Catalog No. AA4234704
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4234704 2 g
AA4234709 10 g
AA4234718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4234704 Supplier Thermo Scientific Chemicals Supplier No. 04234704
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 33685-54-0 | C2H2Cl4 | 169.85 g/mol

1,1,2,2-Tetrachloroethane-d_2 is used as an intermediate in chemical research and in pharmaceutics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33685-54-0
Molecular Formula C2H2Cl4
Molecular Weight (g/mol) 169.85
MDL Number MFCD00037672
InChI Key QPFMBZIOSGYJDE-QDNHWIQGSA-N
Synonym 1,1,2,2-tetrachloroethane-d2, 1,2-dideutero-1,1,2,2-tetrachloroethane, tetrachloro 2 h? ethane, 1,1,2,2-tetrachloroethane-d2 tce-d2, ethane-d2, 1,1,2,2-tetrachloro, ethane-1,2-d2, 1,1,2,2-tetrachloro, 1,1,2,2-tetrachloro-1,2-2h2 ethane, de87d, 1,1,2,2-tetrachloroethane-d2 >99.50 atom % d, 1,1,2,2-tetrachloroethane-d2, ≥99.5 atom % d
PubChem CID 118531
IUPAC Name 1,1,2,2-tetrachloro-1,2-dideuterioethane
SMILES C(C(Cl)Cl)(Cl)Cl

Specifications

Physical Form Liquid
Color Colorless
Quantity 2 g
Density 1.620 g/mL
Refractive Index 1.493
Melting Point -44°C
Boiling Point 147°C
Linear Formula Cl2CDCDCl2
UN Number UN1702
Solubility Information Slightly soluble in water. Soluble in ether,chloroform,ethanol,tetrachloromethane,methanol,acetone,petroleum spirit,carbon disulfide,dimethyl formamide and benzene.
Formula Weight 169.86
Percent Purity 99.6%
Chemical Name or Material 1,1,2,2-Tetrachloroethane-d2, Isotopic
Show More Show Less
Hazard Category H310+H330
Hazard Statement GHS H Statement
H310-H330
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P302+P350-P304+P340-P310-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 1,1,2,2-TETRACHLOROETHANE
EINECSNumber 251-634-1
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.