Learn More
Cambridge Isotope Laboratories 1,1,2,2-Tetrachloroethane-D2 (D, 99.5%) + 0.05% V/V TMS | 33685-54-0 | 10g
Supplier:  Cambridge Isotope Laboratories DLM35TB100
Encompass_Preferred
1,1,2,2-Tetrachloroethane-D2 (D, 99.5%) + 0.05% V/V TMS | Purity: 99.5%% | Mol Wt: 169.86 | 33685-54-0 | 10g
Catalog No. NC1630479