Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2-Tetrachloroethane-d2, for NMR, 99.5+ atom% D

Catalog No. AC321310250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC321310250 25 g
AC321310010 1 g
AC321310050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC321310250 Supplier Thermo Scientific Chemicals Supplier No. 321310250
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 33685-54-0 | C2H2Cl4 | 169.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33685-54-0
Molecular Formula C2H2Cl4
Molecular Weight (g/mol) 169.85
MDL Number MFCD00037672
InChI Key QPFMBZIOSGYJDE-QDNHWIQGSA-N
Synonym 1,1,2,2-tetrachloroethane-d2, 1,2-dideutero-1,1,2,2-tetrachloroethane, tetrachloro 2 h? ethane, 1,1,2,2-tetrachloroethane-d2 tce-d2, ethane-d2, 1,1,2,2-tetrachloro, ethane-1,2-d2, 1,1,2,2-tetrachloro, 1,1,2,2-tetrachloro-1,2-2h2 ethane, de87d, 1,1,2,2-tetrachloroethane-d2 >99.50 atom % d, 1,1,2,2-tetrachloroethane-d2, ≥99.5 atom % d
PubChem CID 118531
IUPAC Name 1,1,2,2-tetrachloro-1,2-dideuterioethane
SMILES C(C(Cl)Cl)(Cl)Cl

Specifications

Physical Form Liquid
Quantity 25 g
Color Colorless
Deuterium (D) 99.50% min.
Density 1.6100g/mL
Linear Formula (Cl2CD)2
Formula Weight 169.86
Grade NMR
Chemical Name or Material 1,1,2,2-Tetrachloroethane-d2, 99.5+ atom% D, for NMR
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if inhaled.
Fatal in contact with skin.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.