Learn More
Medchemexpress LLC 1,1,2,2-tetradeuterio-2-(2-methyl-5-nitroimidazol-1-yl)ethanol | 1261392-47-5 | 98.25% | 5 MG
SDP
Catalog No. 5000406496
Encompass_Preferred
Metronidazole-d4 is a deuterium labeled form of metronidazole, a nitroimidazole antibiotic medication used primarily for anaerobic bacteria and protozoa. This product is for research use only and has not been fully validated for medical applications.
  • Deuterium labeled metronidazole
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-06496 Supplier Medchemexpress LLC Supplier No. HYB0318S15MG
Metronidazole-d4 is a deuterium labeled form of metronidazole, a nitroimidazole antibiotic medication used primarily for anaerobic bacteria and protozoa. This product is for research use only and has not been fully validated for medical applications.
  • Deuterium labeled metronidazole
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs