Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2-Tetrakis(4-methylphenyl)-1,2-ethanediol, TCI America™
SDP

Catalog No. T16521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T16521G Supplier TCI America Supplier No. T16521G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 913-86-0
Molecular Formula C30H30O2
Molecular Weight (g/mol) 422.568
MDL Number MFCD01321188
InChI Key UWVQXJFNNKWNLZ-UHFFFAOYSA-N
Synonym 1,1,2,2-Tetra-p-tolyl-1,2-ethanediol
PubChem CID 136698
IUPAC Name 1,1,2,2-tetrakis(4-methylphenyl)ethane-1,2-diol
SMILES CC1=CC=C(C=C1)C(C2=CC=C(C=C2)C)(C(C3=CC=C(C=C3)C)(C4=CC=C(C=C4)C)O)O

Specifications

Melting Point 177°C
Color White
Quantity 1 g
Formula Weight 422.57
Physical Form Crystalline Powder
Chemical Name or Material 1,1,2,2-Tetrakis(4-methylphenyl)-1,2-ethanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.