Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,1,2,2-Tetraphenyl-1,2-ethanediol

Catalog No. B980710G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B980710G 10 g
B9807100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B980710G Supplier Sigma Aldrich Supplier No. B980710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,1,2,2-Tetraphenyl-1,2-ethanediol

Specifications

CAS 464-72-2
Melting Point 171°C to 173°C (lit.)
Molecular Formula C26H22O2
Linear Formula (C6H5)2 C(OH)C(C6H5)2 OH
MDL Number MFCD00004448
Quantity 10 g
Synonym TPED; Benzopinacol
Molecular Weight (g/mol) 366.45
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.