Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,3-Tetrachloropropane 97.0+%, TCI America™
SDP

Catalog No. T070710ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T070710ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T070710ML Supplier TCI America Supplier No. T070710ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18495-30-2
Molecular Formula C3H4Cl4
Molecular Weight (g/mol) 181.87
MDL Number MFCD00039343
InChI Key BUQMVYQMVLAYRU-UHFFFAOYNA-N
Synonym propane, 1,1,2,3-tetrachloro, propane, tetrachloro, tetrachloropropane, acmc-209em9, buqmvyqmvlayru-uhfffaoysa, propane,1,1,2,3-tetrachloro
PubChem CID 29108
IUPAC Name 1,1,2,3-tetrachloropropane
SMILES ClCC(Cl)C(Cl)Cl

Specifications

Color Yellow
Boiling Point 179°C
Quantity 10 mL
Formula Weight 181.87
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,2,3-Tetrachloropropane
EINECSNumber (2)-0083
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.