Learn More
Newman Sanitary Gasket Company 1 1/2 Inch Silicone Gasket, Tri-Clamp Design, FDA Approved, 1 1/2 in
Catalog No. NC1966703
Encompass_Preferred
This Tri-Clamp Gasket, made from platinum-cured silicone, is ideal for high-purity process applications in pharmaceutical, food, and dairy sectors. Its design ensures superior sealing integrity and broad chemical compatibility across temperatures ranging from -65°F to 450°F. The FDA-approved gasket meets stringent industry standards, setting it apart from competitors by offering reliability and performance for critical processes.
Catalog No. NC1966703 Supplier Newman Sanitary Gasket Company Supplier No. 40RXPX0150
This Tri-Clamp Gasket, made from platinum-cured silicone, is ideal for high-purity process applications in pharmaceutical, food, and dairy sectors. Its design ensures superior sealing integrity and broad chemical compatibility across temperatures ranging from -65°F to 450°F. The FDA-approved gasket meets stringent industry standards, setting it apart from competitors by offering reliability and performance for critical processes.