Learn More
Medchemexpress LLC 1-[1-[2-methoxy-4-(piperidin-4-yl)oxybenzoyl]piperidin-4-yl]-4H-3,1-benzoxazin-2-one | 162045-26-3 | 99.2% | 586.68 g·mol⁻¹ | C33H38N4O6 | 10 MG
SDP
Catalog No. 5000286193
Encompass_Preferred
L-372662 is a research-grade, non-peptide oxytocin receptor antagonist used in pharmacological and receptor-binding studies. It is described as potent and orally active and is supplied as a small-quantity, high-purity research sample suitable for in vitro and in vivo experiments.
  • Potent oxytocin receptor antagonist (reported Ki ≈ 4.8 nM).
  • Orally active in preclinical models.
  • High chemical purity (≈99.2%).
  • Molecular weight 586.68 g·mol⁻¹; formula C33H38N4O6.
  • Provided as a 10 mg research sample for assay development and pharmacology studies.
Catalog No. 50-002-86193 Supplier Medchemexpress LLC Supplier No. HY1501110MG
L-372662 is a research-grade, non-peptide oxytocin receptor antagonist used in pharmacological and receptor-binding studies. It is described as potent and orally active and is supplied as a small-quantity, high-purity research sample suitable for in vitro and in vivo experiments.
  • Potent oxytocin receptor antagonist (reported Ki ≈ 4.8 nM).
  • Orally active in preclinical models.
  • High chemical purity (≈99.2%).
  • Molecular weight 586.68 g·mol⁻¹; formula C33H38N4O6.
  • Provided as a 10 mg research sample for assay development and pharmacology studies.