Learn More
Medchemexpress LLC 1-(1-(2-(piperidin-1-yl)acetyl)piperidin-4-yl)-5-(1-isopropyl-2-oxo-... | 2768712-71-4 | 99.9% | 461.60 g/mol | C27H35N5O2 | 5 MG
SDP
Catalog No. 5000221080
Encompass_Preferred
CDK9-IN-13 is a research-grade small-molecule inhibitor selective for cyclin-dependent kinase 9 (CDK9). It exhibits potent biochemical inhibition of CDK9 (IC50 <3 nM), displays short half-lives in rodents, and is supplied at high purity for biochemical and cellular studies.
  • Potent CDK9 inhibition (IC50 <3 nM)
  • Selective kinase profile with reduced off-target activity
  • High purity (99.94%) suitable for research assays
  • Molecular weight 461.60 g/mol; formula C27H35N5O2
  • Available in small mg quantities for screening and validation
Catalog No. 50-002-21080 Supplier Medchemexpress LLC Supplier No. HY1399805MG
CDK9-IN-13 is a research-grade small-molecule inhibitor selective for cyclin-dependent kinase 9 (CDK9). It exhibits potent biochemical inhibition of CDK9 (IC50 <3 nM), displays short half-lives in rodents, and is supplied at high purity for biochemical and cellular studies.
  • Potent CDK9 inhibition (IC50 <3 nM)
  • Selective kinase profile with reduced off-target activity
  • High purity (99.94%) suitable for research assays
  • Molecular weight 461.60 g/mol; formula C27H35N5O2
  • Available in small mg quantities for screening and validation